Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-8-hydroxy-7-iodoquinoline, 98%

Manufacturer:  Alfa Aesar™ A1113709

Quantity 10g

 View more versions of this product

Catalog No. AAA1113709


  Check Availability
Add to cart

Specifications

Description

Applications
Clioquinol is a metal chelating ligand which displays protective effects in various neurodegenerative states as well as being a component in antifungal and antiprotozoal agents. Clioquinol rescues lead-induced impairments of synaptic plasticity in an animal models

Solubility
Soluble in DMSO (>25 mg/ml), boiling alcohol ((1:43)), methanol, and chloroform ((1:120)).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

130-26-7
305.49
153637
Clioqinol
C9H5ClINO
MFCD00006787
14,5031
5-Chloro-8-hydroxy-7-iodoquinoline

Specifications

≈175°C (dec.)
130-26-7
305.49
153637
Clioqinol
98%
10g
C9H5ClINO
MFCD00006787
14,5031
5-Chloro-8-hydroxy-7-iodoquinoline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H361-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : VC5075000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit