Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-8-hydroxy-7-iodoquinoline 97.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
130-26-7
C9H5ClINO
305.499
MFCD00006787
QCDFBFJGMNKBDO-UHFFFAOYSA-N
clioquinol, iodochlorhydroxyquin, chloroiodoquin, chinoform, chloroiodoquine, 5-chloro-8-hydroxy-7-iodoquinoline, iodochloroxyquinoline, cliquinol, vioform, chlorojodochin
2788
CHEBI:74460
5-chloro-7-iodoquinolin-8-ol
C1=CC2=C(C(=C(C=C2Cl)I)O)N=C1
130-26-7
C9H5ClINO
305.499
MFCD00006787
QCDFBFJGMNKBDO-UHFFFAOYSA-N
clioquinol, iodochlorhydroxyquin, chloroiodoquin, chinoform, chloroiodoquine, 5-chloro-8-hydroxy-7-iodoquinoline, iodochloroxyquinoline, cliquinol, vioform, chlorojodochin
2788
CHEBI:74460
5-chloro-7-iodoquinolin-8-ol
C1=CC2=C(C(=C(C=C2Cl)I)O)N=C1
Quantity 250g
Color Undesignated
Formula Weight 305.50
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material 5-Chloro-8-hydroxy-7-iodoquinoline

RTECSNumber : VC5075000

TSCA : Yes