Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chlorobenzofuroxan, 97%

Click to view available options
Quantity:
25g
17348-69-5
C6H3ClN2O2
170.552
MFCD00068063
DHPQXIQZZCNOLI-UHFFFAOYSA-N
5-chlorobenzofuroxan,5-chlorobenzofurazan 1-oxide,2,1,3-benzoxadiazole, 5-chloro-, monooxide,5-chloro-2,1,3-benzoxadiazol-1-ium-1-olate,5-chloro-2,1,3-benzoxadiazole 1-oxide,acmc-20lz11,5-chloro-beznofurazan oxide,5-chlorobenzfurazan-1-oxide,5-chlorobenzofurazan-1-oxide,5-chlorobenzofurazane 1-oxide
140222
5-chloro-1-oxido-2,1,3-benzoxadiazol-1-ium
C1=CC2=[N+](ON=C2C=C1Cl)[O-]
17348-69-5
C6H3ClN2O2
170.552
MFCD00068063
DHPQXIQZZCNOLI-UHFFFAOYSA-N
5-chlorobenzofuroxan,5-chlorobenzofurazan 1-oxide,2,1,3-benzoxadiazole, 5-chloro-, monooxide,5-chloro-2,1,3-benzoxadiazol-1-ium-1-olate,5-chloro-2,1,3-benzoxadiazole 1-oxide,acmc-20lz11,5-chloro-beznofurazan oxide,5-chlorobenzfurazan-1-oxide,5-chlorobenzofurazan-1-oxide,5-chlorobenzofurazane 1-oxide
140222
5-chloro-1-oxido-2,1,3-benzoxadiazol-1-ium
C1=CC2=[N+](ON=C2C=C1Cl)[O-]
Melting Point 45°C to 48°C
Quantity 25g
Formula Weight 170.56
Percent Purity 97%
Chemical Name or Material 5-Chlorobenzofuroxan

TSCA : No

Recommended Storage : Ambient temperatures