Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloroindole-3-carboxaldehyde, 98%

Click to view available options
Quantity:
1g
5g
827-01-0
C9H6ClNO
179.603
MFCD00175291
YXEXOIGXNYITQH-UHFFFAOYSA-N
5-chloroindole-3-carboxaldehyde,1h-indole-3-carboxaldehyde, 5-chloro,5-chloroindole-3-carbaldehyde,5-chloro-1h-indol-3-carboxaldehyde,pubchem7689,5-chloro-3-formylindole,ksc649s5j,5-chloro indole-3-carboxaldehyde,5-chloro-1h-indole-3-carbaldehyde,5-chloro-1h-indole-3-carboxaldehyde
2795374
5-chloro-1H-indole-3-carbaldehyde
C1=CC2=C(C=C1Cl)C(=CN2)C=O
827-01-0
C9H6ClNO
179.603
MFCD00175291
YXEXOIGXNYITQH-UHFFFAOYSA-N
5-chloroindole-3-carboxaldehyde,1h-indole-3-carboxaldehyde, 5-chloro,5-chloroindole-3-carbaldehyde,5-chloro-1h-indol-3-carboxaldehyde,pubchem7689,5-chloro-3-formylindole,ksc649s5j,5-chloro indole-3-carboxaldehyde,5-chloro-1h-indole-3-carbaldehyde,5-chloro-1h-indole-3-carboxaldehyde
2795374
5-chloro-1H-indole-3-carbaldehyde
C1=CC2=C(C=C1Cl)C(=CN2)C=O
Quantity 5g
Melting Point 210°C to 212°C
Beilstein 123794
Formula Weight 179.61
Percent Purity 98%
Chemical Name or Material 5-Chloroindole-3-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning