Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Chloromethyl)uracil, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
3590-48-5
C5H5ClN2O2
160.56
UCDUBKRXOPMNGH-UHFFFAOYSA-N
5-chloromethyl pyrimidine-2,4 1h,3h-dione,5-chloromethyl-1,3-dihydropyrimidine-2,4-dione,2,4 1h,3h-pyrimidinedione, 5-chloromethyl,5-chloromethyluracil,5-chloromethyl uracil,acmc-1ady4,5-chloromethyl-2,4-pyrimidinediol,5-chloromethyl pyrimidine-2,4-diol,5-chloromethyl uracil 250mg
236125
5-(chloromethyl)-1H-pyrimidine-2,4-dione
C1=C(C(=O)NC(=O)N1)CCl
3590-48-5
C5H5ClN2O2
160.56
UCDUBKRXOPMNGH-UHFFFAOYSA-N
5-chloromethyl pyrimidine-2,4 1h,3h-dione,5-chloromethyl-1,3-dihydropyrimidine-2,4-dione,2,4 1h,3h-pyrimidinedione, 5-chloromethyl,5-chloromethyluracil,5-chloromethyl uracil,acmc-1ady4,5-chloromethyl-2,4-pyrimidinediol,5-chloromethyl pyrimidine-2,4-diol,5-chloromethyl uracil 250mg
236125
5-(chloromethyl)-1H-pyrimidine-2,4-dione
C1=C(C(=O)NC(=O)N1)CCl
Melting Point >350°C
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Formula Weight 160.56
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)uracil

GHS Signal Word: Warning