Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorooxindole, ≥97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
17630-75-0
C8H6ClNO
167.592
MFCD00191927
WWJLCYHYLZZXBE-UHFFFAOYSA-N
5-chlorooxindole,5-chloroindolin-2-one,5-chloro-1,3-dihydro-2h-indol-2-one,5-chloro-2-indolinone,5-chloro-1,3-dihydro-indol-2-one,5-chloro-2-oxindole,2h-indol-2-one, 5-chloro-1,3-dihydro,5-chloro-2,3-dihydro-1h-indol-2-one,2-indolinone, 5-chloro,5-chlorooxindol
152801
5-chloro-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)Cl)NC1=O
17630-75-0
C8H6ClNO
167.592
MFCD00191927
WWJLCYHYLZZXBE-UHFFFAOYSA-N
5-chlorooxindole,5-chloroindolin-2-one,5-chloro-1,3-dihydro-2h-indol-2-one,5-chloro-2-indolinone,5-chloro-1,3-dihydro-indol-2-one,5-chloro-2-oxindole,2h-indol-2-one, 5-chloro-1,3-dihydro,5-chloro-2,3-dihydro-1h-indol-2-one,2-indolinone, 5-chloro,5-chlorooxindol
152801
5-chloro-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)Cl)NC1=O
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Tan
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Beilstein 21, 251
Formula Weight 167.59
Physical Form Powder
Percent Purity ≥97.0%
Chemical Name or Material 5-Chlorooxindole
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Suspected of damaging fertility.

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning