Alfa Aesar™ 5-Chloropentyl acetate, 98%

Manufacturer: Alfa Aesar™ A1111130

Quantity 250g

 View more versions of this product

Catalog No. AAA1111130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20395-28-2
    Molecular Formula CH3CO2(CH2)5Cl
    Formula Weight 164.63
    MDL Number MFCD00013696
    Beilstein 1752107
    Synonym 1-Acetoxy-5-chloropentane
    Chemical Name or Material 5-Chloropentyl acetate

    Specifications

    Formula Weight 164.63
    Molecular Formula CH3CO2(CH2)5Cl
    CAS 20395-28-2
    Beilstein 1752107
    Synonym 1-Acetoxy-5-chloropentane
    Boiling Point 96° to 98°C (11mmHg)
    TSCA TSCA
    MDL Number MFCD00013696
    Flash Point 97°C (206°F)
    Quantity 250g
    Chemical Name or Material 5-Chloropentyl acetate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit