Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chlorothiophene-2-boronic acid, 97%

Click to view available options
Quantity:
1g
162607-18-3
C4H4BClO2S
162.394
MFCD00236030
JFCLNCVCDFUJPO-UHFFFAOYSA-N
5-chlorothiophene-2-boronic acid,5-chloro-2-thienylboronic acid,5-chlorothiophen-2-yl boronic acid,5-chloro-2-thiopheneboronic acid,5-chlorothiophen-2-boronic acid,5-chlorothiophen-2-yl boranediol,5-chlorothiphene 2-boronic acid,5-chloro-2-thienyl boronic acid,boronic acid, 5-chloro-2-thienyl,5-chlorothiophene-2-boronicacid
2734324
(5-chlorothiophen-2-yl)boronic acid
B(C1=CC=C(S1)Cl)(O)O
162607-18-3
C4H4BClO2S
162.394
MFCD00236030
JFCLNCVCDFUJPO-UHFFFAOYSA-N
5-chlorothiophene-2-boronic acid,5-chloro-2-thienylboronic acid,5-chlorothiophen-2-yl boronic acid,5-chloro-2-thiopheneboronic acid,5-chlorothiophen-2-boronic acid,5-chlorothiophen-2-yl boranediol,5-chlorothiphene 2-boronic acid,5-chloro-2-thienyl boronic acid,boronic acid, 5-chloro-2-thienyl,5-chlorothiophene-2-boronicacid
2734324
(5-chlorothiophen-2-yl)boronic acid
B(C1=CC=C(S1)Cl)(O)O
Melting Point ~136°C to 140°C
Quantity 1g
Beilstein 8478804
Formula Weight 162.4
Percent Purity 97%
Chemical Name or Material 5-Chlorothiophene-2-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C