Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorothiophene-2-boronic acid, 97+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
162607-18-3
C4H4BClO2S
162.39
MFCD00236030
JFCLNCVCDFUJPO-UHFFFAOYSA-N
5-chlorothiophene-2-boronic acid, 5-chloro-2-thienylboronic acid, 5-chlorothiophen-2-yl boronic acid, 5-chloro-2-thiopheneboronic acid, 5-chlorothiophen-2-boronic acid, 5-chlorothiophen-2-yl boranediol, 5-chlorothiphene 2-boronic acid, 5-chloro-2-thienyl boronic acid, boronic acid, 5-chloro-2-thienyl, 5-chlorothiophene-2-boronicacid
2734324
(5-chlorothiophen-2-yl)boronic acid
OB(O)C1=CC=C(Cl)S1
162607-18-3
C4H4BClO2S
162.39
MFCD00236030
JFCLNCVCDFUJPO-UHFFFAOYSA-N
5-chlorothiophene-2-boronic acid, 5-chloro-2-thienylboronic acid, 5-chlorothiophen-2-yl boronic acid, 5-chloro-2-thiopheneboronic acid, 5-chlorothiophen-2-boronic acid, 5-chlorothiophen-2-yl boranediol, 5-chlorothiphene 2-boronic acid, 5-chloro-2-thienyl boronic acid, boronic acid, 5-chloro-2-thienyl, 5-chlorothiophene-2-boronicacid
2734324
(5-chlorothiophen-2-yl)boronic acid
OB(O)C1=CC=C(Cl)S1
Melting Point 136.0°C to 140.0°C
Packaging Glass bottle
Quantity 1g
Assay Percent Range 97+%
Solubility Information Solubility in water: insoluble
Formula Weight 162.4
Percent Purity 97+%
Chemical Name or Material 5-Chlorothiophene-2-boronic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning