Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chlorothiophene-2-carboxaldehyde, 97%

Click to view available options
Quantity:
1g
5g
25g
7283-96-7
C5H3ClOS
146.588
MFCD00047090
VWYFITBWBRVBSW-UHFFFAOYSA-N
5-chloro-2-thiophenecarboxaldehyde,2-chloro-5-thiophenecarboxaldehyde,5-chlorothiophene-2-carboxaldehyde,2-thiophenecarboxaldehyde, 5-chloro,5-chloro-2-formylthiophene,2-chloro-5-formylthiophene,5-chlorothiophene-2-aldehyde,5-chloro-2-thiophenecarbaldehyde,5-chloro-2-thiophene-carboxaldehyde,pubchem7742
81700
5-chlorothiophene-2-carbaldehyde
C1=C(SC(=C1)Cl)C=O
7283-96-7
C5H3ClOS
146.588
MFCD00047090
VWYFITBWBRVBSW-UHFFFAOYSA-N
5-chloro-2-thiophenecarboxaldehyde,2-chloro-5-thiophenecarboxaldehyde,5-chlorothiophene-2-carboxaldehyde,2-thiophenecarboxaldehyde, 5-chloro,5-chloro-2-formylthiophene,2-chloro-5-formylthiophene,5-chlorothiophene-2-aldehyde,5-chloro-2-thiophenecarbaldehyde,5-chloro-2-thiophene-carboxaldehyde,pubchem7742
81700
5-chlorothiophene-2-carbaldehyde
C1=C(SC(=C1)Cl)C=O
Boiling Point 88°C to 90°C (9mmHg)
Flash Point 97°C (208°F)
Quantity 5g
Formula Weight 46.6
Percent Purity 97%
Chemical Name or Material 5-Chlorothiophene-2-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C