Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-chlorothiophene-2-carboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
24065-33-6
C5H3ClO2S
162.6
MFCD00041426
QZLSBOVWPHXCLT-UHFFFAOYSA-N
5-chloro-2-thiophenecarboxylic acid,2-thiophenecarboxylic acid, 5-chloro,2-chloro-5-carboxythiophene,5-chloro-2-thiophene carboxylic acid,pubchem7380,maybridge3_003717,acmc-209g8i,rivaroxaban related compound,5-18-06-00177 beilstein handbook reference,ksc204m2j
95048
5-chlorothiophene-2-carboxylic acid
C1=C(SC(=C1)Cl)C(=O)O
24065-33-6
C5H3ClO2S
162.6
MFCD00041426
QZLSBOVWPHXCLT-UHFFFAOYSA-N
5-chloro-2-thiophenecarboxylic acid,2-thiophenecarboxylic acid, 5-chloro,2-chloro-5-carboxythiophene,5-chloro-2-thiophene carboxylic acid,pubchem7380,maybridge3_003717,acmc-209g8i,rivaroxaban related compound,5-18-06-00177 beilstein handbook reference,ksc204m2j
95048
5-chlorothiophene-2-carboxylic acid
C1=C(SC(=C1)Cl)C(=O)O
Melting Point 146°C to 150°C
Color Cream-Yellow
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 162.6
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 5-chlorothiophene-2-carboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning