Alfa Aesar™ 5-Chlorothiophene-2-sulfonyl chloride, 97%

Manufacturer: Alfa Aesar™ A11348-14

Quantity 25g

 View more versions of this product

Catalog No. AAA1134814

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2766-74-7
    Molecular Formula C4H2Cl2O2S2
    Formula Weight 217.09
    MDL Number MFCD00051667
    UN Number UN3265
    Beilstein 130710
    Chemical Name or Material 5-Chlorothiophene-2-sulfonyl chloride

    Specifications

    Formula Weight 217.09
    Molecular Formula C4H2Cl2O2S2
    Health Hazard 1 Danger
    Recommended Storage 2°C to 8°C
    Boiling Point 117° to 120°C (14mmHg)
    CAS 2766-74-7
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    I
    Quantity 25g
    MDL Number MFCD00051667
    UN Number UN3265
    Beilstein 130710
    Flash Point >110°C (230°F)
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 5-Chlorothiophene-2-sulfonyl chloride
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    2°C to 8°C

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    I

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit