Alfa Aesar™ 5-Chlorouracil, 98%

Manufacturer: Alfa Aesar™ A1108422

Quantity 100g

 View more versions of this product

Catalog No. AAA1108422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1820-81-1
    Molecular Formula C4H3ClN2O2
    Formula Weight 146.53
    MDL Number MFCD00006019
    Beilstein 127173
    Synonym 5-Chloro-2,4-dihydroxypyrimidine
    Chemical Name or Material 5-Chlorouracil

    Specifications

    Formula Weight 146.53
    Molecular Formula C4H3ClN2O2
    MDL Number MFCD00006019
    CAS 1820-81-1
    Synonym 5-Chloro-2,4-dihydroxypyrimidine
    RTECS Number YQ9410000
    Quantity 100g
    Beilstein 127173
    Melting Point >300°C
    Chemical Name or Material 5-Chlorouracil
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit