Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyano-2-methylpyridine, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
3222-48-8
C7H6N2
118.14
PBLOYQAQGYUPCM-UHFFFAOYSA-N
5-cyano-2-picoline,5-cyano-2-methylpyridine,6-methylnicotinonitrile,6-methyl-3-pyridinecarbonitrile,3-cyano-6-methylpyridine,3-pyridinecarbonitrile, 6-methyl,pubchem6695,2-methyl-5-cyanopyridine,6-methylnicotinonitrile #,acmc-209z1l
281995
6-methylpyridine-3-carbonitrile
CC1=NC=C(C=C1)C#N
3222-48-8
C7H6N2
118.14
PBLOYQAQGYUPCM-UHFFFAOYSA-N
5-cyano-2-picoline,5-cyano-2-methylpyridine,6-methylnicotinonitrile,6-methyl-3-pyridinecarbonitrile,3-cyano-6-methylpyridine,3-pyridinecarbonitrile, 6-methyl,pubchem6695,2-methyl-5-cyanopyridine,6-methylnicotinonitrile #,acmc-209z1l
281995
6-methylpyridine-3-carbonitrile
CC1=NC=C(C=C1)C#N
Melting Point 81°C to 86°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 118.14
Percent Purity 97%
Chemical Name or Material 5-Cyano-2-methylpyridine

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger