Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyanothiophene-2-boronic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
305832-67-1
C5H4BNO2S
152.97
MFCD02094029
ZEOMEPSYIIQIND-UHFFFAOYSA-N
5-cyanothiophene-2-boronic acid,5-cyanothiophen-2-yl boronic acid,5-cyanothiophen-2-yl-2-boronic acid,5-boronothiophene-2-carbonitrile,5-cyano-2-thienyl boronic acid,5-dihydroxyboranyl thiophene-2-carbonitrile,5-cyano-2-thienylboronic acid,boronic acid, 5-cyano-2-thienyl,5-cyanothiophen-2-boronic acid,acmc-1ctts
4198743
(5-cyanothiophen-2-yl)boronic acid
B(C1=CC=C(S1)C#N)(O)O
305832-67-1
C5H4BNO2S
152.97
MFCD02094029
ZEOMEPSYIIQIND-UHFFFAOYSA-N
5-cyanothiophene-2-boronic acid,5-cyanothiophen-2-yl boronic acid,5-cyanothiophen-2-yl-2-boronic acid,5-boronothiophene-2-carbonitrile,5-cyano-2-thienyl boronic acid,5-dihydroxyboranyl thiophene-2-carbonitrile,5-cyano-2-thienylboronic acid,boronic acid, 5-cyano-2-thienyl,5-cyanothiophen-2-boronic acid,acmc-1ctts
4198743
(5-cyanothiophen-2-yl)boronic acid
B(C1=CC=C(S1)C#N)(O)O
Melting Point 175°C to 180°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Color Pink to Beige
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and thf
Formula Weight 152.97
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 5-Cyanothiophene-2-boronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning