Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Cyanothiophene-2-carboxaldehyde, 95%

Click to view available options
Quantity:
250mg
1g
5g
21512-16-3
C6H3NOS
137.156
MFCD09037804
PZIFYWVUYHMYOA-UHFFFAOYSA-N
5-cyano-2-thiophene carbaldehyde,5-cyano-2-thiophenecarboxaldehyde,5-cyanothiophene-2-carboxaldehyde,2-thiophenecarbonitrile, 5-formyl,5-cyano-2-thiophenecarbaldehyde,2-formyl-5-cyanothiophen,2-cyano-5-formylthiophene,2-formyl-5-cyanothiophene,2-cyano-5-formyl-thiophene,5-cyanothiophene-2-aldehyde
12280004
5-formylthiophene-2-carbonitrile
C1=C(SC(=C1)C#N)C=O
21512-16-3
C6H3NOS
137.156
MFCD09037804
PZIFYWVUYHMYOA-UHFFFAOYSA-N
5-cyano-2-thiophene carbaldehyde,5-cyano-2-thiophenecarboxaldehyde,5-cyanothiophene-2-carboxaldehyde,2-thiophenecarbonitrile, 5-formyl,5-cyano-2-thiophenecarbaldehyde,2-formyl-5-cyanothiophen,2-cyano-5-formylthiophene,2-formyl-5-cyanothiophene,2-cyano-5-formyl-thiophene,5-cyanothiophene-2-aldehyde
12280004
5-formylthiophene-2-carbonitrile
C1=C(SC(=C1)C#N)C=O
Applications
5-Cyanothiophene-2-carboxaldehyde is found in caper and may have pest control activity against meloidogyne incognita.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
Melting Point 90°C to 95°C
Color White
UN Number UN3439
Beilstein 115517
Formula Weight 137.16
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Cyanothiophene-2-carboxaldehyde

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.