Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Diethylamino-2-pentanol, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
5412-69-1
C9H21NO
159.273
MFCD00004559
OAQYRNDEOJQVBN-UHFFFAOYSA-N
5-diethylamino-2-pentanol,5-diethylamino pentan-2-ol,2-pentanol, 5-diethylamino,2-pentanol,5-diethylamino,oaqyrndeojqvbn-uhfffaoysa
95388
5-(diethylamino)pentan-2-ol
CCN(CC)CCCC(C)O
5412-69-1
C9H21NO
159.273
MFCD00004559
OAQYRNDEOJQVBN-UHFFFAOYSA-N
5-diethylamino-2-pentanol,5-diethylamino pentan-2-ol,2-pentanol, 5-diethylamino,2-pentanol,5-diethylamino,oaqyrndeojqvbn-uhfffaoysa
95388
5-(diethylamino)pentan-2-ol
CCN(CC)CCCC(C)O
Density 0.8600g/mL
Color Colorless to Yellow
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4440 to 1.4460 (20°C, 589nm)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula (C2H5)2N(CH2)3CH(OH)CH3
Formula Weight 159.27
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 5-Diethylamino-2-pentanol, 96%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-497-6

Recommended Storage : Product may darken during storage