Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Ethyl-2-methylpyridine 98%, ACROS Organics™

Click to view available options
Quantity:
250g
Packaging:
Glass bottle
104-90-5
C8H11N
121.183
MFCD00006344
NTSLROIKFLNUIJ-UHFFFAOYSA-N
aldehydine,5-ethyl-2-picoline,2-methyl-5-ethylpyridine,aldehydkollidin,aldehydecollidine,pyridine, 5-ethyl-2-methyl,2,5-aldehydine,2-picoline, 5-ethyl,3-ethyl-6-methylpyridine,6-methyl-3-ethylpyridine
7728
5-ethyl-2-methylpyridine
CCC1=CN=C(C=C1)C
104-90-5
C8H11N
121.183
MFCD00006344
NTSLROIKFLNUIJ-UHFFFAOYSA-N
aldehydine,5-ethyl-2-picoline,2-methyl-5-ethylpyridine,aldehydkollidin,aldehydecollidine,pyridine, 5-ethyl-2-methyl,2,5-aldehydine,2-picoline, 5-ethyl,3-ethyl-6-methylpyridine,6-methyl-3-ethylpyridine
7728
5-ethyl-2-methylpyridine
CCC1=CN=C(C=C1)C
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Water 0.1% max. (K.F.)
Packaging Glass bottle
Density 0.9190g/mL
Infrared Spectrum Authentic
Quantity 250g
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C2H5C5H3NCH3
Beilstein 20, 248
Merck Index 15, 3896
Formula Weight 121.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 5-Ethyl-2-methylpyridine, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-250-0

RTECSNumber : TJ6825000

TSCA : TSCA

Recommended Storage : product darkens with time