Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Ethyl-2-thiophenecarboxaldehyde, 98%

Click to view available options
Quantity:
1g
5g
25g
36880-33-8
C7H8OS
140.2
MFCD00143457
CLQXZICUPGZTPE-UHFFFAOYSA-N
5-ethyl-2-thiophenecarboxaldehyde,5-ethyl-2-thiophenecarbaldehyde,2-thiophenecarboxaldehyde, 5-ethyl,5-ethyl-2-thiophene carboxaldehyde,2-ethyl-5-formylthiophene,5-ethyl-thiophene-2-carbaldehyde,acmc-1csxu,ksc497m6n,bidd:gt0099,5-ethylthiophene-2-carboxaldehyde
169830
5-ethylthiophene-2-carbaldehyde
CCC1=CC=C(S1)C=O
36880-33-8
C7H8OS
140.2
MFCD00143457
CLQXZICUPGZTPE-UHFFFAOYSA-N
5-ethyl-2-thiophenecarboxaldehyde,5-ethyl-2-thiophenecarbaldehyde,2-thiophenecarboxaldehyde, 5-ethyl,5-ethyl-2-thiophene carboxaldehyde,2-ethyl-5-formylthiophene,5-ethyl-thiophene-2-carbaldehyde,acmc-1csxu,ksc497m6n,bidd:gt0099,5-ethylthiophene-2-carboxaldehyde
169830
5-ethylthiophene-2-carbaldehyde
CCC1=CC=C(S1)C=O
Quantity 1g
Flash Point 109°C (228°F)
Boiling Point 65°C (2mmHg)
Beilstein 108539
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 5-Ethyl-2-thiophenecarboxaldehyde