Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 5-Ethyl-2-thiophenecarboxaldehyde, 98%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Density | 1.12 |
Flash Point | 109°C (228°F) |
Refractive Index | 1.57 |
Sensitivity | Air Sensitive |
Boiling Point | 65°C (2mmHg) |
Quantity | 25g |
Beilstein | 108539 |
Formula Weight | 140.21 |
Percent Purity | 98% |
Chemical Name or Material | 5-Ethyl-2-thiophenecarboxaldehyde |
Safety and Handling
EINECSNumber : 253-252-0
TSCA : Yes
Recommended Storage : Ambient temperatures