Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Ethylcyclohexane-1,3-dione hemihydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
57641-76-6
C8H12O2
140.182
MFCD06202339
VPRYSOJVBSTQKE-UHFFFAOYSA-N
2734798
5-ethylcyclohexane-1,3-dione
CCC1CC(=O)CC(=O)C1
57641-76-6
C8H12O2
140.182
MFCD06202339
VPRYSOJVBSTQKE-UHFFFAOYSA-N
2734798
5-ethylcyclohexane-1,3-dione
CCC1CC(=O)CC(=O)C1
Packaging Glass bottle
Color Beige
Water 5% max.
Quantity 1g
Infrared Spectrum Authentic
Melting Point 49°C to 53°C
Assay Percent Range 98%
Formula Weight 149.19
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 5-Ethylcyclohexane-1, 3-dione hemihydrate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat, drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.