Learn More
Aat BioquestSupplier Diversity Partner 5-FAM Azide DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 131
5-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group. 5-FAM is the purified single isomer of carboxyfluorescein.
Catalog No. 50-195-2739