Learn More
Aat BioquestSupplier Diversity Partner 5-FAM, SE [5-Carboxyfluorescein, succinimidyl ester] *CAS 92557-80-7* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 114
5-FAM, SE is the amine-reactive succinimidyl ester of single isomer 5-FAM acid. It is one of the most popular green fluorescent reagents used for labeling peptides, proteins and nucleotides.
Catalog No. 50-195-2723