Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotano 5 Finger Premium Leather Primate Catch Glove™

Click to view available options
Hand:
Left
Right
Specific
Quantity:
10 Pairs/Cs.
15 Pairs/Cs.
20 Gloves/Cs.
25 Pairs/Cs.
30 Gloves/Cs.
50 Gloves/Cs.

Premium grade suede split leather gloves offer protection against bites, scratches, and lacerations when handling Macaque-sized primates. Designed for trained professionals working in Lab Animal Research (LAR) facilities.

  • Protects against bites, scratches, and lacerations when working with Macaque-sized primates
  • Premium grade suede leather for dexterity, flexibility, and bite protection
  • Full grain hide on the sleeve provides scratch resistance
  • Fleeced poly cotton liner for added insulation, sweat absorption, and comfort
  • High strength seam construction for additional strength and durability
  • Non-exposed stitching prevents snagging
  • Exceeds test method EN388: 2016 + A1:2018 6.5 Puncture Resistance levels
  • Made in the USA
Certifications/Compliance (Europe) EN 388:2016 + A1:2018 6.5 Puncture Resistance: Performance Level 4 (Highest Level)
Cuff Material Full Grain, Scratch Resistant Hide
Color Gray
Grade Premium
Hand Left
Latex Free Yes
Material Suede Leather
Palm Thickness (Metric) 1.4 to 1.6 mm
Product Type Animal Handling Gloves
Size Universal
For Use With (Application) Protection Against Bites, Scratches, Punctures, and Lacerations When Handling Macaque-Sized Primates
Glove Cuff Color Black
Glove Cuff Style Gauntlet
Liner Material Fleeced Polycotton
Quantity 20 Gloves/Cs.
Series NBCG-OFA
Thumb Style Keystone
Length (English) 25.5 in.
Show More Show Less