Alfa Aesar™ 5-Fluoro-2,4-dichloropyrimidine, 98%

Manufacturer: Alfa Aesar™ H2628803

Quantity 1g

 View more versions of this product

Catalog No. AAH2628803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2927-71-1
    Molecular Formula C4HCl2FN2
    Formula Weight 166.97
    MDL Number MFCD00233551
    UN Number UN3261
    Chemical Name or Material 5-Fluoro-2,4-dichloropyrimidine

    Specifications

    Formula Weight 166.97
    CAS 2927-71-1
    UN Number UN3261
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Boiling Point 80°C (16mmHg)
    Melting Point 36° to 39°C
    Quantity 1g
    Molecular Formula C4HCl2FN2
    MDL Number MFCD00233551
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Flash Point 106°C (223°F)
    Chemical Name or Material 5-Fluoro-2,4-dichloropyrimidine
    Assay 98%

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit