Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoro-2-hydroxy-3-nitropyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
136888-20-5
C5H3FN2O3
158.088
MFCD05662412
BLFUHTOZIOQGBU-UHFFFAOYSA-N
5-fluoro-2-hydroxy-3-nitropyridine,5-fluoro-3-nitropyridin-2-ol,5-fluoro-3-nitropyridin-2 1h-one,2-hydroxy-3-nitro-5-fluoropyridine,2-hydroxy-5-fluoro-3-nitropyridine,2 1h-pyridinone, 5-fluoro-3-nitro,pubchem2975,acmc-1cgzr,ksc520k3n,5-fluoro-2-hydroxy-3-nitrop
10583047
5-fluoro-3-nitro-1H-pyridin-2-one
C1=C(C(=O)NC=C1F)[N+](=O)[O-]
136888-20-5
C5H3FN2O3
158.088
MFCD05662412
BLFUHTOZIOQGBU-UHFFFAOYSA-N
5-fluoro-2-hydroxy-3-nitropyridine,5-fluoro-3-nitropyridin-2-ol,5-fluoro-3-nitropyridin-2 1h-one,2-hydroxy-3-nitro-5-fluoropyridine,2-hydroxy-5-fluoro-3-nitropyridine,2 1h-pyridinone, 5-fluoro-3-nitro,pubchem2975,acmc-1cgzr,ksc520k3n,5-fluoro-2-hydroxy-3-nitrop
10583047
5-fluoro-3-nitro-1H-pyridin-2-one
C1=C(C(=O)NC=C1F)[N+](=O)[O-]
Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 250mg
Beilstein 7582013
Formula Weight 158.09
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-hydroxy-3-nitropyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning