Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-hydroxypyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
51173-05-8
C5H4FNO
113.09
MFCD03092918
KLULSYPVWLJZAO-UHFFFAOYSA-N
5-fluoro-2-hydroxypyridine,5-fluoropyridin-2-ol,2-hydroxy-5-fluoropyridine,5-fluoropyridin-2 1h-one,3-fluoro-6-hydroxypyridine,2 1h-pyridinone, 5-fluoro,5-fluoro-2 1h-pyridinone,5-fluoro-2-pyridinol,5-fluoropyridin-2-ol hydrobromide,pubchem6566
2762864
5-fluoro-1H-pyridin-2-one
C1=CC(=O)NC=C1F
51173-05-8
C5H4FNO
113.09
MFCD03092918
KLULSYPVWLJZAO-UHFFFAOYSA-N
5-fluoro-2-hydroxypyridine,5-fluoropyridin-2-ol,2-hydroxy-5-fluoropyridine,5-fluoropyridin-2 1h-one,3-fluoro-6-hydroxypyridine,2 1h-pyridinone, 5-fluoro,5-fluoro-2 1h-pyridinone,5-fluoro-2-pyridinol,5-fluoropyridin-2-ol hydrobromide,pubchem6566
2762864
5-fluoro-1H-pyridin-2-one
C1=CC(=O)NC=C1F
Infrared Spectrum Authentic
Color White
Melting Point 152.0°C to 153.0°C
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 113.09
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-hydroxypyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
If medical advice is needed,have product container or label at hand.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning