Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5'-Fluoro-2'-iodoacetophenone, 97%

Click to view available options
Quantity:
250mg
1g
5g
914225-70-0
C8H6FIO
264.038
MFCD09033159
NRLSRIONJVBZDT-UHFFFAOYSA-N
5'-fluoro-2'-iodoacetophenone,1-5-fluoro-2-iodophenyl ethanone,2'-iodo-5'-fluoroacetophenone,1-5-fluoro-2-iodophenyl ethan-1-one,1-5-fluoro-2-iodo-phenyl ethanone,5-fluoro-2-iodoacetophenone,pubchem15742,5-fluoro-2-iodoacetophenone,1-5-fluoro-2-iodo-phenyl-ethanone,1-5-fluoranyl-2-iodanyl-phenyl ethanone
29934881
1-(5-fluoro-2-iodophenyl)ethanone
CC(=O)C1=C(C=CC(=C1)F)I
914225-70-0
C8H6FIO
264.038
MFCD09033159
NRLSRIONJVBZDT-UHFFFAOYSA-N
5'-fluoro-2'-iodoacetophenone,1-5-fluoro-2-iodophenyl ethanone,2'-iodo-5'-fluoroacetophenone,1-5-fluoro-2-iodophenyl ethan-1-one,1-5-fluoro-2-iodo-phenyl ethanone,5-fluoro-2-iodoacetophenone,pubchem15742,5-fluoro-2-iodoacetophenone,1-5-fluoro-2-iodo-phenyl-ethanone,1-5-fluoranyl-2-iodanyl-phenyl ethanone
29934881
1-(5-fluoro-2-iodophenyl)ethanone
CC(=O)C1=C(C=CC(=C1)F)I
Applications
5′-Fluoro-2′-iodoacetophenone is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Incompatible with light and oxidizing agents.
Quantity 1g
Color Brown
Sensitivity Light Sensitive
Beilstein 22044599
Formula Weight 264.04
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 5'-Fluoro-2'-iodoacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning