Alfa Aesar™ 5-Fluoro-2-methoxybenzoic acid, 97%

Manufacturer: Alfa Aesar™ H3238906

Quantity 5g

 View more versions of this product

Catalog No. AAH3238906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 394-04-7
    Molecular Formula C8H7FO3
    Formula Weight 170.14
    MDL Number MFCD00671765
    Chemical Name or Material 5-Fluoro-2-methoxybenzoic acid

    Specifications

    Formula Weight 170.14
    MDL Number MFCD00671765
    CAS 394-04-7
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Molecular Formula C8H7FO3
    Health Hazard 1 Warning
    Melting Point 86° to 90°C
    Quantity 5g
    Chemical Name or Material 5-Fluoro-2-methoxybenzoic acid
    Assay 97%

    Description

    Applications
    5-Fluoro-3-hydroxy-2-methylbenzoic acid 1 was synthesized from 5-fluoro-2-methylbenzoic acid by nitration, reduction, diazotization and hydrolysis. Another methylcarbamoyl-substituted benzylamine 12 was synthesized from 5-fluoro-2-methylbenzoic acid via the sequential bromination,azido replacement and the reduction.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit