Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoro-2-methoxybenzonitrile, 98+%

Click to view available options
Quantity:
1g
5g
189628-38-4
C8H6FNO
151.14
MFCD02683503
XNCVSNKXADJGOG-UHFFFAOYSA-N
2-cyano-4-fluoroanisole,3-fluoro-6-methoxybenzonitrile,benzonitrile, 5-fluoro-2-methoxy,5-fluoro-2-methoxybenzenecarbonitrile,pubchem3502,acmc-1bxaq,xncvsnkxadjgog-uhfffaoysa,benzonitrile,5-fluoro-2-methoxy,buttpark 80\01-23,5-fluoro-2-methyloxy benzonitrile
2782746
5-fluoro-2-methoxybenzonitrile
COC1=C(C=C(C=C1)F)C#N
189628-38-4
C8H6FNO
151.14
MFCD02683503
XNCVSNKXADJGOG-UHFFFAOYSA-N
2-cyano-4-fluoroanisole,3-fluoro-6-methoxybenzonitrile,benzonitrile, 5-fluoro-2-methoxy,5-fluoro-2-methoxybenzenecarbonitrile,pubchem3502,acmc-1bxaq,xncvsnkxadjgog-uhfffaoysa,benzonitrile,5-fluoro-2-methoxy,buttpark 80\01-23,5-fluoro-2-methyloxy benzonitrile
2782746
5-fluoro-2-methoxybenzonitrile
COC1=C(C=C(C=C1)F)C#N
Melting Point 115°C to 118°C
Quantity 1g
UN Number UN3439
Formula Weight 151.14
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2-methoxybenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.