Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoro-2-methoxypyridine-3-carboxaldehyde, 98%

Click to view available options
Quantity:
250mg
1g
5g
351410-62-3
C7H6FNO2
155.128
MFCD04972398
YAGPZRLCMRMSFP-UHFFFAOYSA-N
5-fluoro-2-methoxynicotinaldehyde,5-fluoro-3-formyl-2-methoxypyridine,3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy,5-fluoro-2-methoxy-3-pyridinecarboxaldehyde,2-methoxy-5-fluoropyridine-3-carbaldehyde,5-fluoro-2-methoxy-pyridine-3-carbaldehyde,5-fluoro-2-methoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde,5-fluoro-2-methoxy,5-fluoro-2-methoxy-3-pyridine carboxaldehyde,pubchem16100
22678323
5-fluoro-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(C=C1C=O)F
351410-62-3
C7H6FNO2
155.128
MFCD04972398
YAGPZRLCMRMSFP-UHFFFAOYSA-N
5-fluoro-2-methoxynicotinaldehyde,5-fluoro-3-formyl-2-methoxypyridine,3-pyridinecarboxaldehyde, 5-fluoro-2-methoxy,5-fluoro-2-methoxy-3-pyridinecarboxaldehyde,2-methoxy-5-fluoropyridine-3-carbaldehyde,5-fluoro-2-methoxy-pyridine-3-carbaldehyde,5-fluoro-2-methoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde,5-fluoro-2-methoxy,5-fluoro-2-methoxy-3-pyridine carboxaldehyde,pubchem16100
22678323
5-fluoro-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(C=C1C=O)F
Sensitivity Air & Moisture Sensitive
Quantity 1g
Beilstein 14158282
Formula Weight 155.13
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methoxypyridine-3-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C