Alfa Aesar™ 5-Fluoro-2-methoxypyridine, 97%

Manufacturer: Alfa Aesar™ H3099103

Quantity 1g

 View more versions of this product

Catalog No. AAH3099103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51173-04-7
    Molecular Formula C6H6FNO
    Formula Weight 127.12
    MDL Number MFCD03095252
    UN Number UN1993
    Chemical Name or Material 5-Fluoro-2-methoxypyridine

    Specifications

    Formula Weight 127.12
    UN Number UN1993
    Recommended Storage −30° to −10°C
    Molecular Formula C6H6FNO
    MDL Number MFCD03095252
    CAS 51173-04-7
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Quantity 1g
    Chemical Name or Material 5-Fluoro-2-methoxypyridine
    Assay 97%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    −30° to −10°C

    GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit