Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (5-Fluoro-2-methyl-3-indolyl)acetic acid, 97%

Click to view available options
Quantity:
1g
5g
71987-67-2
C11H10FNO2
207.204
MFCD02664389
VJZAMNBVIWOUJR-UHFFFAOYSA-N
5-fluoro-2-methyl-1h-indol-3-yl acetic acid,5-fluoro-2-methyl-1h-indol-3-yl-acetic acid,2-5-fluoro-2-methyl-1h-indol-3-yl acetic acid,1h-indole-3-acetic acid, 5-fluoro-2-methyl,1h-indole-3-aceticacid, 5-fluoro-2-methyl,2-5-fluoro-2-methylindol-3-yl acetic acid,chembl82614,2-methyl-5-fluoro-1h-indole-3-acetic acid
2772338
2-(5-fluoro-2-methyl-1H-indol-3-yl)acetic acid
CC1=C(C2=C(N1)C=CC(=C2)F)CC(=O)O
71987-67-2
C11H10FNO2
207.204
MFCD02664389
VJZAMNBVIWOUJR-UHFFFAOYSA-N
5-fluoro-2-methyl-1h-indol-3-yl acetic acid,5-fluoro-2-methyl-1h-indol-3-yl-acetic acid,2-5-fluoro-2-methyl-1h-indol-3-yl acetic acid,1h-indole-3-acetic acid, 5-fluoro-2-methyl,1h-indole-3-aceticacid, 5-fluoro-2-methyl,2-5-fluoro-2-methylindol-3-yl acetic acid,chembl82614,2-methyl-5-fluoro-1h-indole-3-acetic acid
2772338
2-(5-fluoro-2-methyl-1H-indol-3-yl)acetic acid
CC1=C(C2=C(N1)C=CC(=C2)F)CC(=O)O
Quantity 5g
Beilstein 399422
Formula Weight 207.2
Physical Form Solid
Percent Purity 97%
Chemical Name or Material (5-Fluoro-2-methyl-3-indolyl)acetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning