Alfa Aesar™ 5-Fluoro-2-methylbenzyl alcohol, 98%

Manufacturer: Alfa Aesar™ H3264803

Quantity 1g

 View more versions of this product

Catalog No. AAH3264803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22062-54-0
    Molecular Formula C8H9FO
    Formula Weight 140.16
    MDL Number MFCD01631527
    Chemical Name or Material 5-Fluoro-2-methylbenzyl alcohol

    Specifications

    Formula Weight 140.16
    Molecular Formula C8H9FO
    MDL Number MFCD01631527
    CAS 22062-54-0
    Melting Point 36° to 40°C
    Quantity 1g
    Chemical Name or Material 5-Fluoro-2-methylbenzyl alcohol
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit