Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoro-2-methylindole, 95%

Click to view available options
Quantity:
1g
5g
399-72-4
C9H8FN
149.168
MFCD02093649
JJIUISYYTFDATN-UHFFFAOYSA-N
5-fluoro-2-methylindole,1h-indole, 5-fluoro-2-methyl,zlchem 457,pubchem2158,2-methyl-5-fluoroindole,5-fluoro-2-methyl indole,5-fluoro-2-methyl-indole,ksc222g8n,5-fluoro-2-methylindole,5-fluoro-2-methylindole
2778715
5-fluoro-2-methyl-1H-indole
CC1=CC2=C(N1)C=CC(=C2)F
399-72-4
C9H8FN
149.168
MFCD02093649
JJIUISYYTFDATN-UHFFFAOYSA-N
5-fluoro-2-methylindole,1h-indole, 5-fluoro-2-methyl,zlchem 457,pubchem2158,2-methyl-5-fluoroindole,5-fluoro-2-methyl indole,5-fluoro-2-methyl-indole,ksc222g8n,5-fluoro-2-methylindole,5-fluoro-2-methylindole
2778715
5-fluoro-2-methyl-1H-indole
CC1=CC2=C(N1)C=CC(=C2)F
Quantity 1g
Melting Point 98°C to 101°C
Beilstein 116627
Formula Weight 149.17
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Fluoro-2-methylindole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning