Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoro-2-methylphenol, 98%

Manufacturer:  Alfa Aesar™ B2560603

Quantity 1g

 View more versions of this product

Catalog No. AAB2560603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

452-85-7
126.13
UN3265
5-Fluoro-o-cresol; 4-Fluoro-2-hydroxytoluene
C7H7FO
MFCD00673009
3234863
5-Fluoro-2-methylphenol

Specifications

Phenol-like
1g
C7H7FO
MFCD00673009
3234863
5-Fluoro-2-methylphenol
66°C (4mmHg)
452-85-7
126.13
UN3265
5-Fluoro-o-cresol; 4-Fluoro-2-hydroxytoluene
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit