Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoro-2-nitrobenzoic acid, 97+%

Click to view available options
Quantity:
1g
5g
320-98-9
C7H4FNO4
185.11
MFCD00055635
GHYZIXDKAPMFCS-UHFFFAOYSA-N
5-fluoro-2-nitrobenzoic acid,2-nitro-5-fluorobenzoic acid,3-fluoro-6-nitrobenzoic acid,5-fluoro-2-nitro-benzoic acid,benzoic acid, 5-fluoro-2-nitro,2-carboxy-4-fluoronitrobenzene,pubchem1309,acmc-1aiv3,5-fluoro-2nitrobenzoic acid,5-fluor-2-nitrobenzoic acid
2737420
C1=CC(=C(C=C1F)C(=O)O)[N+](=O)[O-]
320-98-9
C7H4FNO4
185.11
MFCD00055635
GHYZIXDKAPMFCS-UHFFFAOYSA-N
5-fluoro-2-nitrobenzoic acid,2-nitro-5-fluorobenzoic acid,3-fluoro-6-nitrobenzoic acid,5-fluoro-2-nitro-benzoic acid,benzoic acid, 5-fluoro-2-nitro,2-carboxy-4-fluoronitrobenzene,pubchem1309,acmc-1aiv3,5-fluoro-2nitrobenzoic acid,5-fluor-2-nitrobenzoic acid
2737420
C1=CC(=C(C=C1F)C(=O)O)[N+](=O)[O-]
Quantity 5g
Melting Point 134°C to 135°C
Formula Weight 185.11
Percent Purity ≥97%
Chemical Name or Material 5-Fluoro-2-nitrobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning