Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-nitrotoluene 96%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
446-33-3
C7H6FNO2
155.128
MFCD00007283
JHFOWEGCZWLHNW-UHFFFAOYSA-N
5-fluoro-2-nitrotoluene,3-fluoro-6-nitrotoluene,4-fluoro-2-methylnitrobenzene,2-nitro-5-fluoro toluene,benzene, 4-fluoro-2-methyl-1-nitro,toluene, 5-fluoro-2-nitro,2-nitro-5-fluorotoluene,2-methyl-4-fluoronitrobenzene,3-methyl-4-nitrofluorobenzene,4-fluoro-2-methyl-1-nitro-benzene
94956
4-fluoro-2-methyl-1-nitrobenzene
CC1=C(C=CC(=C1)F)[N+](=O)[O-]
446-33-3
C7H6FNO2
155.128
MFCD00007283
JHFOWEGCZWLHNW-UHFFFAOYSA-N
5-fluoro-2-nitrotoluene,3-fluoro-6-nitrotoluene,4-fluoro-2-methylnitrobenzene,2-nitro-5-fluoro toluene,benzene, 4-fluoro-2-methyl-1-nitro,toluene, 5-fluoro-2-nitro,2-nitro-5-fluorotoluene,2-methyl-4-fluoronitrobenzene,3-methyl-4-nitrofluorobenzene,4-fluoro-2-methyl-1-nitro-benzene
94956
4-fluoro-2-methyl-1-nitrobenzene
CC1=C(C=CC(=C1)F)[N+](=O)[O-]
Refractive Index 1.5260 to 1.5300 (20°C, 589nm)
Quantity 50g
Packaging Glass bottle
Color Green-Yellow to Yellow
Density 1.2700g/mL
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula CH3C6H3(NO2)F
Beilstein 05, II, 250
Formula Weight 155.13
Physical Form Liquid After Melting
Percent Purity ≥95%
Chemical Name or Material 5-Fluoro-2-nitrotoluene, 96%
Show More Show Less

GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-164-4