Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoro-2-nitrotoluene, 98+%

Click to view available options
Quantity:
10g
50g
446-33-3
C7H6FNO2
155.128
MFCD00007283
JHFOWEGCZWLHNW-UHFFFAOYSA-N
5-fluoro-2-nitrotoluene, 3-fluoro-6-nitrotoluene, 4-fluoro-2-methylnitrobenzene, 2-nitro-5-fluoro toluene, benzene, 4-fluoro-2-methyl-1-nitro, toluene, 5-fluoro-2-nitro, 2-nitro-5-fluorotoluene, 2-methyl-4-fluoronitrobenzene, 3-methyl-4-nitrofluorobenzene, 4-fluoro-2-methyl-1-nitro-benzene
94956
4-fluoro-2-methyl-1-nitrobenzene
CC1=C(C=CC(=C1)F)[N+](=O)[O-]
446-33-3
C7H6FNO2
155.128
MFCD00007283
JHFOWEGCZWLHNW-UHFFFAOYSA-N
5-fluoro-2-nitrotoluene, 3-fluoro-6-nitrotoluene, 4-fluoro-2-methylnitrobenzene, 2-nitro-5-fluoro toluene, benzene, 4-fluoro-2-methyl-1-nitro, toluene, 5-fluoro-2-nitro, 2-nitro-5-fluorotoluene, 2-methyl-4-fluoronitrobenzene, 3-methyl-4-nitrofluorobenzene, 4-fluoro-2-methyl-1-nitro-benzene
94956
4-fluoro-2-methyl-1-nitrobenzene
CC1=C(C=CC(=C1)F)[N+](=O)[O-]
Density 1.27
Refractive Index 1.528
Flash Point 87°C (188°F)
Melting Point ~20°C
Quantity 10g
Boiling Point 97°C to 98°C (10mmHg)
UN Number UN2810
Beilstein 2046652
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2-nitrotoluene
Show More Show Less

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 207-164-4

TSCA : No

Recommended Storage : Ambient temperatures