Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-oxindole, 97%, ACROS Organics™

Click to view available options
Quantity:
500mg
56341-41-4
C8H6FNO
151.14
DDIIYGHHUMKDGI-UHFFFAOYSA-N
5-fluorooxindole,5-fluoroindolin-2-one,5-fluoro-2-oxindole,5-fluoro-1,3-dihydro-2h-indol-2-one,2h-indol-2-one, 5-fluoro-1,3-dihydro,5-fluoro-1,3-dihydro-indol-2-one,5-fluoro-2-indolinone,5-fluoro-2,3-dihydro-1h-indol-2-one,5-fluoro indoline-2-one,5-fluoro-2,3-dihydro-2-oxoindole
3731012
5-fluoro-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)F)NC1=O
56341-41-4
C8H6FNO
151.14
DDIIYGHHUMKDGI-UHFFFAOYSA-N
5-fluorooxindole,5-fluoroindolin-2-one,5-fluoro-2-oxindole,5-fluoro-1,3-dihydro-2h-indol-2-one,2h-indol-2-one, 5-fluoro-1,3-dihydro,5-fluoro-1,3-dihydro-indol-2-one,5-fluoro-2-indolinone,5-fluoro-2,3-dihydro-1h-indol-2-one,5-fluoro indoline-2-one,5-fluoro-2,3-dihydro-2-oxoindole
3731012
5-fluoro-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)F)NC1=O
Melting Point 144°C to 146°C
Color Brown to White
Infrared Spectrum Authentic
Quantity 500mg
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 151.14
Physical Form Powder or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-oxindole
Show More Show Less