Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoro-2-(trifluoromethyl)aniline, 97%

Manufacturer:  Alfa Aesar™ H2665703

Quantity 1g

 View more versions of this product

Catalog No. AAH2665703


  Check Availability
Add to cart

Specifications

Description

Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

827-20-3
179.12
UN2811
5-Fluoro-2-(trifluoromethyl)aniline
C7H5F4N
MFCD06660353
2-Amino-4-fluorobenzotrifluoride

Specifications

Characteristic
1g
C7H5F4N
MFCD06660353
2-Amino-4-fluorobenzotrifluoride
97%
29° to 32°C
827-20-3
179.12
UN2811
5-Fluoro-2-(trifluoromethyl)aniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit