Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-DL-tryptophan, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
154-08-5
C11H11FN2O2
222.219
INPQIVHQSQUEAJ-UHFFFAOYSA-N
5-fluoro-dl-tryptophan,5-fluorotryptophan,dl-5-fluorotryptophan,2-amino-3-5-fluoro-1h-indol-3-yl propanoic acid,tryptophan, 5-fluoro,tryptophan, 5-fluoro-, dl,5-fluoro-d,l-tryptophan,2-amino-3-5-fluoroindol-3-yl propanoic acid,h-dl-trp 5-f-oh,dl-tryptophan, 5-fluoro
9577
CHEBI:77837
2-amino-3-(5-fluoro-1H-indol-3-yl)propanoic acid
C1=CC2=C(C=C1F)C(=CN2)CC(C(=O)O)N
154-08-5
C11H11FN2O2
222.219
INPQIVHQSQUEAJ-UHFFFAOYSA-N
5-fluoro-dl-tryptophan,5-fluorotryptophan,dl-5-fluorotryptophan,2-amino-3-5-fluoro-1h-indol-3-yl propanoic acid,tryptophan, 5-fluoro,tryptophan, 5-fluoro-, dl,5-fluoro-d,l-tryptophan,2-amino-3-5-fluoroindol-3-yl propanoic acid,h-dl-trp 5-f-oh,dl-tryptophan, 5-fluoro
9577
CHEBI:77837
2-amino-3-(5-fluoro-1H-indol-3-yl)propanoic acid
C1=CC2=C(C=C1F)C(=CN2)CC(C(=O)O)N
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Beige
Assay Percent Range 99+%
Formula Weight 222.22
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material 5-Fluoro-DL-tryptophan