Learn More
5-Fluorocytosine Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259147
Catalog No. NBP259147
Add to cart
Description
Description
5-Fluorocytosine Polyclonal antibody specifically detects 5-Fluorocytosine in Human, Mouse samples. It is validated for ELISA, Immunoprecipitation.
Specifications
Specifications
5-Fluorocytosine
Unconjugated
Serum with 0.05% Sodium Azide
5-fC
The exact immunogen is propietary information.
Whole Antiserum
RUO
Primary
Human, Mouse
ELISA, Immunoprecipitation
ELISA 1:10000, Immunoprecipitation
Antiserum
Rabbit
IgG
100 ul
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal