Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Fluoroindirubinoxime, Tocris Bioscience™

Catalog Number 403350
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
861214-33-7
C16H10FN3O2
295.27
MFCD18086923
LBUXOYSDWHGJNP-UHFFFAOYSA-N
5'-fluoroindirubinoxime, 5'-fluoro-1h,1'h-2,3' biindolylidene-3,2'dione 3-oxime, 5/'-fluoroindirubinoxime, d0t9fe, a05-a4b1-ii, 3z-5-fluoro-3-3e-3-hydroxyiminoindolin-2-ylidene indolin-2-one, 5-fluoro-3-3-hydroxyimino-1,3-dihydro-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
11580208
5'-fluoro-3-(hydroxyamino)-1H,2'H-[2,3'-biindol]-2'-one
ONC1=C(NC2=CC=CC=C12)C1=C2C=C(F)C=CC2=NC1=O
This item is not returnable. View return policy
861214-33-7
C16H10FN3O2
295.27
MFCD18086923
LBUXOYSDWHGJNP-UHFFFAOYSA-N
5'-fluoroindirubinoxime, 5'-fluoro-1h,1'h-2,3' biindolylidene-3,2'dione 3-oxime, 5/'-fluoroindirubinoxime, d0t9fe, a05-a4b1-ii, 3z-5-fluoro-3-3e-3-hydroxyiminoindolin-2-ylidene indolin-2-one, 5-fluoro-3-3-hydroxyimino-1,3-dihydro-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
11580208
5'-fluoro-3-(hydroxyamino)-1H,2'H-[2,3'-biindol]-2'-one
ONC1=C(NC2=CC=CC=C12)C1=C2C=C(F)C=CC2=NC1=O

FLT3 inhibitor; displays antiproliferative activity

Inhibitor of FMS-like receptor tyrosine kinase-3 (FLT3) (IC50 = 15 nM). Displays selectivity for FLT3 against 6 other kinases including EGFR. Displays antiproliferative activity against the MV4;11 cell line (expressing constitutively active FLT3) and a number of cancer cell lines, including SNU-638 (stomach carcinoma) and HT-1080 (fibrosarcoma).
Quantity 50 mg
Formula Weight 295.27
Percent Purity >99%
Chemical Name or Material 5'-Fluoroindirubinoxime
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit