Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroindole-3-carboxaldehyde, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
2338-71-8
C9H6FNO
163.15
YUAJKGBLPVLADK-UHFFFAOYSA-N
5-fluoroindole-3-carboxaldehyde,5-fluoro-1h-indole-3-carboxaldehyde,5-fluoroindole-3-carbaldehyde,1h-indole-3-carboxaldehyde, 5-fluoro,3-formyl-5-fluoroindole,pubchem7267,zlchem 1197,5-fluoro-3-formylindole,5-fluoroindole-3-aldehyde,acmc-209g2v
259089
5-fluoro-1H-indole-3-carbaldehyde
C1=CC2=C(C=C1F)C(=CN2)C=O
2338-71-8
C9H6FNO
163.15
YUAJKGBLPVLADK-UHFFFAOYSA-N
5-fluoroindole-3-carboxaldehyde,5-fluoro-1h-indole-3-carboxaldehyde,5-fluoroindole-3-carbaldehyde,1h-indole-3-carboxaldehyde, 5-fluoro,3-formyl-5-fluoroindole,pubchem7267,zlchem 1197,5-fluoro-3-formylindole,5-fluoroindole-3-aldehyde,acmc-209g2v
259089
5-fluoro-1H-indole-3-carbaldehyde
C1=CC2=C(C=C1F)C(=CN2)C=O
Melting Point 170°C to 172°C
Quantity 5g
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 163.15
Percent Purity 99%
Chemical Name or Material 5-Fluoroindole-3-carboxaldehyde

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning