Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Fluoroindole, 99%

Click to view available options
Quantity:
1g
5g
25g
399-52-0
C8H6FN
135.141
MFCD00005671
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
5-fluoroindole,1h-indole, 5-fluoro,5-fluoro indole,indole, 5-fluoro,5-fluoro-indole,5-fluor-1h-indol,5-fluoroindol,5-fluoro-indol,3fuf,pubchem1681
67861
CHEBI:72818
5-fluoro-1H-indole
C1=CC2=C(C=CN2)C=C1F
399-52-0
C8H6FN
135.141
MFCD00005671
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
5-fluoroindole,1h-indole, 5-fluoro,5-fluoro indole,indole, 5-fluoro,5-fluoro-indole,5-fluor-1h-indol,5-fluoroindol,5-fluoro-indol,3fuf,pubchem1681
67861
CHEBI:72818
5-fluoro-1H-indole
C1=CC2=C(C=CN2)C=C1F
Quantity 5g
Melting Point 42°C to 47°C
Flash Point >110°C (230°F)
Beilstein 112350
Formula Weight 135.14
Percent Purity 99%
Chemical Name or Material 5-Fluoroindole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-917-4

TSCA : No

Recommended Storage : Keep cold