Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroisatin, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
443-69-6
C8H4FNO2
165.123
MFCD00022795
GKODDAXOSGGARJ-UHFFFAOYSA-N
5-fluoroisatin,5-fluoroindoline-2,3-dione,5-fluoro isatin,5-fluoro-2,3-dihydro-1h-indole-2,3-dione,5-fluoroisatine,5-fluoro-2,3-indoledione,5-fluoroindole-2,3-dione,1h-indole-2,3-dione, 5-fluoro,5-fluoro-2,3-indolinedione,5-fluoro-2,3-indolindione
236566
5-fluoro-1H-indole-2,3-dione
C1=CC2=C(C=C1F)C(=O)C(=O)N2
443-69-6
C8H4FNO2
165.123
MFCD00022795
GKODDAXOSGGARJ-UHFFFAOYSA-N
5-fluoroisatin,5-fluoroindoline-2,3-dione,5-fluoro isatin,5-fluoro-2,3-dihydro-1h-indole-2,3-dione,5-fluoroisatine,5-fluoro-2,3-indoledione,5-fluoroindole-2,3-dione,1h-indole-2,3-dione, 5-fluoro,5-fluoro-2,3-indolinedione,5-fluoro-2,3-indolindione
236566
5-fluoro-1H-indole-2,3-dione
C1=CC2=C(C=C1F)C(=O)C(=O)N2
Melting Point 224°C to 228°C
Quantity 1g
Color Red
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 165.12
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 5-Fluoroisatin, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.