Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluororesorcinol, TCI America™

Click to view available options
Quantity:
200mg
1g
75996-29-1
C6H5FO2
128.102
MFCD08457182
LYFBZGKZAZBANN-UHFFFAOYSA-N
12658778
5-fluorobenzene-1,3-diol
C1=C(C=C(C=C1O)F)O
75996-29-1
C6H5FO2
128.102
MFCD08457182
LYFBZGKZAZBANN-UHFFFAOYSA-N
12658778
5-fluorobenzene-1,3-diol
C1=C(C=C(C=C1O)F)O
Color Undesignated
Quantity 200mg
Melting Point 138°C
Formula Weight 128.10
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 5-Fluororesorcinol