Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-(Fmoc-amino)valeric acid, 98%

Click to view available options
Quantity:
1g
5g
25g
123622-48-0
C20H21NO4
339.391
MFCD00235889
ULLSWWGYZWBPHK-UHFFFAOYSA-N
fmoc-5-ava-oh,5-9h-fluoren-9-yl methoxy carbonyl amino pentanoic acid,5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid,5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid,pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino,ambotzfaa1651,n-fmoc-5-aminopentanoic acid,5-fmoc-amino pentanoic acid,5-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid,fmoc-5-ava-oh fmoc-5-aminopentanoic acid
2756098
5-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(=O)O
123622-48-0
C20H21NO4
339.391
MFCD00235889
ULLSWWGYZWBPHK-UHFFFAOYSA-N
fmoc-5-ava-oh,5-9h-fluoren-9-yl methoxy carbonyl amino pentanoic acid,5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid,5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid,pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino,ambotzfaa1651,n-fmoc-5-aminopentanoic acid,5-fmoc-amino pentanoic acid,5-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid,fmoc-5-ava-oh fmoc-5-aminopentanoic acid
2756098
5-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(=O)O
Applications
5-(Fmoc-amino)valeric acid is used as pharmaceutical intermediate.

Notes
Protect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with
Color White
Quantity 25g
Beilstein 3565625
Formula Weight 339.39
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 5-(Fmoc-amino)valeric acid

storageNote1 : 2° to 8°C