Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Formamido-1-(2-formyloxyethyl)-1H-pyrazole, 97%

Click to view available options
Quantity:
1g
5g
116856-18-9
C7H9N3O3
183.167
MFCD09744040
ADNBFDFIEWMUBJ-UHFFFAOYSA-N
2-5-formamido-1h-pyrazol-1-yl ethyl formate,5-formamide-1-2-formyloxyethl pyrazole,5-formamido-1-2-formyloxyethyl pyrazole,2-5-formamidopyrazol-1-yl ethyl formate,5-formylamino-1-2-formyloxyethyl pyrazole,5-formamido-1-2-formyloxy ethyl pyrazole,pubchem15369,tpc-i141,5-formamido-l-2-formyloxyethyl pyrazole,5-formamido-1-2-formyloxyethyl pyrazole
14996567
2-(5-formamidopyrazol-1-yl)ethyl formate
C1=C(N(N=C1)CCOC=O)NC=O
116856-18-9
C7H9N3O3
183.167
MFCD09744040
ADNBFDFIEWMUBJ-UHFFFAOYSA-N
2-5-formamido-1h-pyrazol-1-yl ethyl formate,5-formamide-1-2-formyloxyethl pyrazole,5-formamido-1-2-formyloxyethyl pyrazole,2-5-formamidopyrazol-1-yl ethyl formate,5-formylamino-1-2-formyloxyethyl pyrazole,5-formamido-1-2-formyloxy ethyl pyrazole,pubchem15369,tpc-i141,5-formamido-l-2-formyloxyethyl pyrazole,5-formamido-1-2-formyloxyethyl pyrazole
14996567
2-(5-formamidopyrazol-1-yl)ethyl formate
C1=C(N(N=C1)CCOC=O)NC=O
Quantity 5g
Beilstein 7762121
Formula Weight 183.17
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-Formamido-1-(2-formyloxyethyl)-1H-pyrazole